Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human HVEM/TNFRSF14 Alexa Fluor 405-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB356V100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB356V100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB356V100 Supplier R&D Systems Supplier No. FAB356V100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

HVEM/TNFRSF14 Monoclonal specifically detects HVEM/TNFRSF14 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen HVEM/TNFRSF14
Applications Flow Cytometry
Classification Monoclonal
Clone 94801
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 405
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. AAB58354
Gene Alias ATAR, CD270, CD270 antigen, CD40-like protein, Herpes virus entry mediator A, Herpesvirus entry mediator A, HveA, HVEMTR2HVEAATAR, LIGHTR, TNFRSF14, tumor necrosis factor receptor superfamily member 14, tumor necrosis factor receptor superfamily, member 14 (herpesvirus entrymediator), Tumor necrosis factor receptor-like 2, tumor necrosis factor receptor-like gene2
Gene Symbols TNFRSF14
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human HVEM/TNFRSF14 Pro37-Val202 (Ser108Thr and Ala140Arg) Accession # AAB58354
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8764
Test Specificity Detects human HVEM/TNFRSF14 in direct ELISAs and Western blots. In direct ELISAs, approximately 5% cross-reactivity with recombinant human (rh) CD30 and no cross-reactivity with rh4-1BB, rhCD27, rhCD40, rhDR3, rhDR6, rhFas, rhGITR, rhNGF-R, rhOPG, rhRANK, recombinant mouse (rm) RANK, rhTAJ, rhTNF RI, rhTNF RII, or rmTNF RI is observed.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.