Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human HVEM/TNFRSF14 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB356SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB356SP 25 μg
MAB356100 100 μg
MAB356500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB356SP Supplier R&D Systems Supplier No. MAB356SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 4 publications

HVEM/TNFRSF14 Monoclonal specifically detects HVEM/TNFRSF14 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready, ELISA Capture (Matched Antibody Pair).

Specifications

Antigen HVEM/TNFRSF14
Applications Western Blot, Flow Cytometry, CyTOF, ELISA
Classification Monoclonal
Clone 94801
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. AAB58354
Gene Alias ATAR, CD270, CD270 antigen, CD40-like protein, Herpes virus entry mediator A, Herpesvirus entry mediator A, HveA, HVEMTR2HVEAATAR, LIGHTR, TNFRSF14, tumor necrosis factor receptor superfamily member 14, tumor necrosis factor receptor superfamily, member 14 (herpesvirus entrymediator), Tumor necrosis factor receptor-like 2, tumor necrosis factor receptor-like gene2
Gene Symbols TNFRSF14
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human HVEM/TNFRSF14 Pro37-Val202 (Ser108Thr and Ala140Arg) Accession # AAB58354
Purification Method Protein A or G purified from ascites
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8764
Test Specificity Detects human HVEM/TNFRSF14 in direct ELISAs and Western blots. In direct ELISAs, approximately 5% cross-reactivity with recombinant human (rh) CD30 and no cross-reactivity with rh4-1BB, rhCD27, rhCD40, rhDR3, rhDR6, rhFas, rhGITR, rhNGF-R, rhOPG, rhRANK, recombinant mouse (rm) RANK, rhTAJ, rhTNF RI, rhTNF RII, or rmTNF RI is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.