Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human HVEM/TNFRSF14 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB3561SP
This item is not returnable.
View return policy
Description
HVEM/TNFRSF14 Monoclonal specifically detects HVEM/TNFRSF14 in Human samples. It is validated for Immunohistochemistry.Specifications
| HVEM/TNFRSF14 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ATAR, CD270, CD270 antigen, CD40-like protein, Herpes virus entry mediator A, Herpesvirus entry mediator A, HveA, HVEMTR2HVEAATAR, LIGHTR, TNFRSF14, tumor necrosis factor receptor superfamily member 14, tumor necrosis factor receptor superfamily, member 14 (herpesvirus entrymediator), Tumor necrosis factor receptor-like 2, tumor necrosis factor receptor-like gene2 | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 8764 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Immunohistochemistry | |
| 94804 | |
| Immunohistochemistry 8-25 ug/mL | |
| Q92956.3 | |
| TNFRSF14 | |
| Mouse myeloma cell line NS0-derived recombinant human HVEM/TNFRSF14 Pro37-Val202 (predicted) Accession # Q92956.3 | |
| 25 μg | |
| Primary | |
| Detects human HVEM/TNFRSF14 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human (rh) 4-1BB, rhBAFF R, rhDR3, rhDR6, rhCD27, rhCD30, rhCD40, rhEDAR, rhFas, rhGITR, rhLT beta R, rhNGF R, rhOPG, recombinant mouse OX40, rhRANK, rhTAJ, rhTNF RI or rhTNF R2 is observed. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction