Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human HVEM/TNFRSF14 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
$200.50 - $491.00
Specifications
Antigen | HVEM/TNFRSF14 |
---|---|
Clone | 2742C |
Dilution | Immunohistochemistry 3-25 ug/mL |
Applications | Immunohistochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB3562SP
![]() |
R&D Systems
MAB3562SP |
25 μg |
Each for $200.50
|
|
|||||
MAB3562100
![]() |
R&D Systems
MAB3562100 |
100 μg |
Each for $491.00
|
|
|||||
Description
HVEM/TNFRSF14 Monoclonal antibody specifically detects HVEM/TNFRSF14 in Human samples. It is validated for ImmunohistochemistrySpecifications
HVEM/TNFRSF14 | |
Immunohistochemistry 3-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
ATAR, CD270, CD270 antigen, CD40-like protein, Herpes virus entry mediator A, Herpesvirus entry mediator A, HveA, HVEMTR2HVEAATAR, LIGHTR, TNFRSF14, tumor necrosis factor receptor superfamily member 14, tumor necrosis factor receptor superfamily, member 14 (herpesvirus entrymediator), Tumor necrosis factor receptor-like 2, tumor necrosis factor receptor-like gene2 | |
Human embryonic kidney cell HEK293-derived human HVEM/TNFRSF14, Pro37-Val202, Accession # Q92956 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
2742C | |
Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Human | |
Q92956 | |
8764 | |
IgG | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title