Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human HVEM/TNFRSF14 APC-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 3 publications
Supplier: R&D Systems FAB356A
This item is not returnable.
View return policy
Description
HVEM/TNFRSF14 Monoclonal specifically detects HVEM/TNFRSF14 in Human samples. It is validated for Flow Cytometry.Specifications
HVEM/TNFRSF14 | |
Monoclonal | |
APC | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
ATAR, CD270, CD270 antigen, CD40-like protein, Herpes virus entry mediator A, Herpesvirus entry mediator A, HveA, HVEMTR2HVEAATAR, LIGHTR, TNFRSF14, tumor necrosis factor receptor superfamily member 14, tumor necrosis factor receptor superfamily, member 14 (herpesvirus entrymediator), Tumor necrosis factor receptor-like 2, tumor necrosis factor receptor-like gene2 | |
Mouse | |
Protein A or G purified from ascites | |
RUO | |
8764 | |
Human | |
Purified |
Flow Cytometry | |
94801 | |
Flow Cytometry 10 uL/10^6 cells | |
AAB58354 | |
TNFRSF14 | |
Mouse myeloma cell line NS0-derived recombinant human HVEM/TNFRSF14 Pro37-Val202 (Ser108Thr and Ala140Arg) Accession # AAB58354 | |
100 Tests | |
Primary | |
Detects human HVEM/TNFRSF14 in direct ELISAs and Western blots. In direct ELISAs, approximately 5% cross-reactivity with recombinant human (rh) CD30 and no cross-reactivity with rh4-1BB, rhCD27, rhCD40, rhDR3, rhDR6, rhFas, rhGITR, rhNGF-R, rhOPG, rhRANK, recombinant mouse (rm) RANK, rhTAJ, rhTNF RI, rhTNF RII, or rmTNF RI is observed. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction