Learn More
Description
Specifications
Specifications
| Quantity | 1 Kit (for 15 Plates) |
| Species | Human |
| Conjugate | Biotin |
| Assay Range | 12.50 to 800 pg/mL |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 3373 |
| Synonym | EC 3.2.1.35, FUS2, HYAL1, HYAL-1, hyaluronidase 1, hyaluronoglucosaminidase 1, Hyaluronoglucosaminidase-1, luCa-1, LUCA1, LUCA1hyaluronidase-1, Lung carcinoma protein 1, MGC45987, NAT6, plasma hyaluronidase, tumor suppressor LUCA-1 |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
