Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IBSP/Sialoprotein II Antibody, R&D Systems™

Catalog No. AF4014
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF4014 100 μg
AF4014SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF4014 Supplier R&D Systems Supplier No. AF4014
This item is not returnable. View return policy

Goat Polyclonal Antibody

IBSP/Sialoprotein II Polyclonal specifically detects IBSP/Sialoprotein II in Human samples. It is validated for Western Blot.

Specifications

Antigen IBSP/Sialoprotein II
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL
Gene Accession No. AAC95490
Gene Alias BNSP, Bone sialoprotein, Bone sialoprotein 2, BSP, BSP 2, BSP II, BSP2, BSPII, BSP-II, Cell binding sialoprotein, IBSP, Integrin binding sialoprotein, SP II, SPII, SP-II
Gene Symbols IBSP
Host Species Goat
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human IBSP/Sialoprotein II Phe17-Gln317 Accession # AAC95490
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3381
Test Specificity Detects human IBSP/Sialoprotein II in direct ELISAs and Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.