Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human ICAM-2/CD102 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF244SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF244SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF244SP Supplier R&D Systems Supplier No. AF244SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 3 publications

ICAM-2/CD102 Polyclonal specifically detects ICAM-2/CD102 in Human samples. It is validated for Western Blot, Flow Cytometry, Adhesion Blockade, CyTOF-ready.

Specifications

Antigen ICAM-2/CD102
Applications Western Blot, Flow Cytometry, Adhesion Blockade, CyTOF
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Adhesion Blockade, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. CAA33630
Gene Alias CD102, CD102 antigen, ICAM2, ICAM-2, intercellular adhesion molecule 2
Gene Symbols ICAM2
Host Species Goat
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human ICAM-2/CD102 Lys25-Gln223 Accession # CAA33630
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3384
Test Specificity Detects human ICAM-2/CD102 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) ICAM-1 and rhICAM-3 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.