Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals Human ICAM-2/CD102 ELISA Kit (Colorimetric)
SDP

Catalog No. NB158625 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB158625 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB158625 Supplier Novus Biologicals Supplier No. NBP314631
Add to Cart
Add to Cart
This item is not returnable. View return policy

Specifications

Assay Range 0.75-24 U/ml
Assay Sensitivity 0.2 U/ml
Product Type ELISA Kit (Colorimetric)
Sample Type Cell Culture Supernatant, Plasma, Serum
Specificity Recognizes both natural and recombinant human soluble ICAM-2/CD102
For Use With (Application) ELISA
Gene ID (Entrez) 3384
Gene Symbol ICAM2
Quantity 1 Kit
Research Discipline Adaptive Immunity, Immunology
Storage Requirements Store at 4C. Do not freeze.
Synonym CD102, CD102 antigen, ICAM-2, intercellular adhesion molecule 2
Target Human
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.