Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human ICAM-3/CD50 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $667.00
Specifications
Antigen | ICAM-3/CD50 |
---|---|
Clone | 76205 |
Dilution | Western Blot 1 ug/mL, Adhesion Blockade |
Applications | Western Blot, Adhesion Blockade |
Classification | Monoclonal |
Description
ICAM-3/CD50 Monoclonal specifically detects ICAM-3/CD50 in Human samples. It is validated for Western Blot, Adhesion Blockade.Specifications
ICAM-3/CD50 | |
Western Blot 1 ug/mL, Adhesion Blockade | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CD50, CD50 antigen, CDW50CDw50, ICAM3, ICAM-3, ICAM-Rintercellular adhesion molecule-3, intercellular adhesion molecule 3 | |
ICAM3 | |
IgG1 | |
Protein A or G purified from ascites | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
76205 | |
Western Blot, Adhesion Blockade | |
Unconjugated | |
Mouse | |
Human | |
CAA49473 | |
3385 | |
Mouse myeloma cell line NS0-derived recombinant human ICAM-3/CD50 Gln30-His485 Accession # CAA49473 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human ICAM-3/CD50 in direct ELISAs and Western blots. Does not cross-react with recombinant human (rh) ICAM-1 or rhICAM-2. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title