Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human ICAM-3/CD50 Antibody, R&D Systems™

Catalog No. BBA2901M
Change view
Click to view available options
Quantity:
1 mg
200 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BBA2901M 1 mg
BBA29200 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. BBA2901M Supplier R&D Systems Supplier No. BBA2901M
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 1 publication

ICAM-3/CD50 Monoclonal specifically detects ICAM-3/CD50 in Human samples. It is validated for Western Blot, Flow Cytometry, Adhesion Blockade, CyTOF-ready.

Specifications

Antigen ICAM-3/CD50
Applications CyTOF, Western Blot
Classification Monoclonal
Clone Cal 3.10
Conjugate Unconjugated
Formulation Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Host Species Mouse
Immunogen Recombinant human ICAM-3/CD50 Fc Chimera
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3385
Test Specificity Detects human ICAM-3/CD50 in Western blots. Does not cross-react with recombinant human (rh) ICAM-1 or rhVCAM-1.
Reconstitution Sterile PBS to a final concentration of 0.5mg/mL.
Target Species Human
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.