Learn More
Human ICAM-3/CD50 Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | ICAM-3/CD50 |
| Applications | Western Blot, Flow Cytometry, CyTOF |
| Classification | Polyclonal |
| Concentration | LYOPH |
| Conjugate | Unconjugated |
| Dilution | Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready |
| Gene Alias | CD50, CD50 antigen, CDW50CDw50, ICAM3, ICAM-3, ICAM-Rintercellular adhesion molecule-3, intercellular adhesion molecule 3 |
| Gene Symbols | ICAM3 |
| Host Species | Sheep |
| Immunogen | Chinese hamster ovary cell line CHO-derived recombinant human ICAM-3/CD50 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.