Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human ICAM-3/CD50 Antibody, R&D Systems™

Goat Polyclonal Antibody
$200.50 - $549.50
Specifications
Antigen | ICAM-3/CD50 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready |
Applications | Western Blot, Flow Cytometry, CyTOF |
Classification | Polyclonal |
Description
ICAM-3/CD50 Polyclonal specifically detects ICAM-3/CD50 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.Specifications
ICAM-3/CD50 | |
Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
Polyclonal | |
Goat | |
Human | |
CD50, CD50 antigen, CDW50CDw50, ICAM3, ICAM-3, ICAM-Rintercellular adhesion molecule-3, intercellular adhesion molecule 3 | |
ICAM3 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Flow Cytometry, CyTOF | |
Unconjugated | |
RUO | |
CAA49473 | |
3385 | |
Mouse myeloma cell line NS0-derived recombinant human ICAM-3/CD50 Gln30-His485 Accession # CAA49473 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human ICAM-3/CD50 in direct ELISAs and Western blots. In Western blots, approximately 20% cross-reactivity with recombinant human ICAM-1 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title