Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human ICAM-4 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF7179
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF7179 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF7179 Supplier R&D Systems Supplier No. AF7179
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

ICAM-4 Polyclonal specifically detects ICAM-4 in Human samples. It is validated for Western Blot, Simple Western, Flow Cytometry, CyTOF-ready.

Specifications

Antigen ICAM-4
Applications Western Blot, Flow Cytometry, CyTOF
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.2 ug/mL, Simple Western 10 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready
Gene Accession No. Q14773
Gene Alias CD242, CD242 antigen, ICAM4, ICAM-4, intercellular adhesion molecule 4, intercellular adhesion molecule 4 (Landsteiner-Wiener blood group), intercellular adhesion molecule 4 (LW blood group), intercellular adhesion molecule 4, Landsteiner-Wiener blood group, Landsteiner-Wiener blood group, Landsteiner-Wiener blood group antigen a, Landsteiner-Wiener blood group glycoprotein, LWLW blood group protein
Gene Symbols ICAM4
Host Species Sheep
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human ICAM-4 Ala31-Ala240 Accession # Q14773
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3386
Test Specificity Detects human ICAM-4 in direct ELISAs and Western blots.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.