Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human ICAM-5 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB1950
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1950 100 μg
MAB1950SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB1950 Supplier R&D Systems Supplier No. MAB1950
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

ICAM-5 Monoclonal specifically detects ICAM-5 in Human samples. It is validated for ELISA Capture (Matched Antibody Pair).

Specifications

Antigen ICAM-5
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 283607
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9UMF0
Gene Alias ICAM5, ICAM-5, intercellular adhesion molecule 5, intercellular adhesion molecule 5, telencephalin, Telencephalin, TLCNTelencephalin, TLNtelencephalin
Gene Symbols ICAM5
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human ICAM-5 Ala28-Glu570 Accession # Q9UMF0
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7087
Test Specificity Detects human ICAM-5 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human (rh) CD31, rhCEACAM-1, recombinant mouse (rm) DCC, rhICAM-1, -2, -3, rmICAM-5, rhMAdCAM-1, or rmVCAM-1 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.