Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human ICAM-5 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF1950
This item is not returnable.
View return policy
Description
ICAM-5 Polyclonal specifically detects ICAM-5 in Human samples. It is validated for Western Blot, Simple Western.Specifications
ICAM-5 | |
Polyclonal | |
Unconjugated | |
Q9UMF0 | |
ICAM5 | |
Mouse myeloma cell line NS0-derived recombinant human ICAM-5 Ala28-Glu570 Accession # Q9UMF0 | |
100 μg | |
Primary | |
Detects human ICAM-5 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 10% cross-reactivity with recombinant mouse ICAM-5 is observed and less than 2% cross-reactivity with recombinant human (rh) ICAM-1, rhICAM-2 and rhICAM-3 is observed. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL, Simple Western 1 ug/mL | |
ICAM5, ICAM-5, intercellular adhesion molecule 5, intercellular adhesion molecule 5, telencephalin, Telencephalin, TLCNTelencephalin, TLNtelencephalin | |
Goat | |
Affinity Purified | |
RUO | |
7087 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction