Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human ICAT Antibody, R&D Systems™

Goat Polyclonal Antibody
$200.50 - $526.50
Specifications
| Antigen | ICAT/CTNNBIP1 |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
ICAT/CTNNBIP1 Polyclonal specifically detects ICAT/CTNNBIP1 in Human samples. It is validated for Western Blot.Specifications
| ICAT/CTNNBIP1 | |
| Western Blot 0.1 ug/mL | |
| Polyclonal | |
| Goat | |
| Human | |
| beta-catenin-interacting protein ICAT, catenin, beta interacting protein 1, CTNNBIP1, ICATbeta-interacting protein 1, Inhibitor of beta-catenin and Tcf-4 | |
| CTNNBIP1 | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Q9NSA3 | |
| 56998 | |
| E. coli-derived recombinant human ICAT Met1-Gln81 Accession # Q9NSA3 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects human ICAT in direct ELISAs and Western blots. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title