Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human ICOS Antibody, R&D Systems™

Mouse Monoclonal Antibody
$208.00 - $582.50
Specifications
Antigen | ICOS |
---|---|
Clone | 669214 |
Dilution | Flow Cytometry 0.25 ug/10^6 cells, Blockade of Receptor-ligand Interaction |
Applications | Flow Cytometry, Blockade of Receptor-ligand Interaction |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB69753SP
![]() |
R&D Systems
MAB69753SP |
25 μg |
Each for $208.00
|
|
|||||
MAB69753100
![]() |
R&D Systems
MAB69753100 |
100 μg |
Each for $582.50
|
|
|||||
Description
ICOS Monoclonal specifically detects ICOS in Human samples. It is validated for Flow Cytometry, Blockade of Receptor-ligand Interaction.Specifications
ICOS | |
Flow Cytometry 0.25 ug/10^6 cells, Blockade of Receptor-ligand Interaction | |
Monoclonal | |
Supernatant | |
Human | |
Q9Y6W8 | |
29851 | |
IgG3 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
669214 | |
Flow Cytometry, Blockade of Receptor-ligand Interaction | |
Unconjugated | |
Mouse | |
Lyophilized from a 0.2 μm filtered solution in PBS and NaCl with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
Activation-inducible lymphocyte immunomediatory molecule, AILIM, AILIMCD278, CD278, CD278 antigen, CRP-1, CVID1, inducible costimulator, inducible T-cell costimulator, inducible T-cell co-stimulator, MGC39850 | |
NS0 mouse myeloma cell line transfected with human ICOS and mouse myeloma cell line NS0-derived recombinant human ICOS cocktail Glu21-Phe141 Accession # Q9Y6W8 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human ICOS in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title