Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IDO2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB10098100
This item is not returnable.
View return policy
Description
IDO2 Monoclonal specifically detects IDO2 in Human samples. It is validated for Immunohistochemistry.Specifications
| IDO2 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| EC 1.13.11.-, IDO-2, INDOL1, INDOL1indoleamine-pyrrole 2,3 dioxygenase-like 1, indoleamine 2,3-dioxygenase 2, Indoleamine 2,3-dioxygenase-2, indoleamine 2,3-dioxygenase-like 1 protein, Indoleamine 2,3-dioxygenase-like protein 1, Indoleamine-pyrrole 2,3-dioxygenase-like protein 1 | |
| Synthetic peptide containing human IDO2, Met1-Val407, Accession # Q6ZQW0 | |
| 100 μg | |
| 169355 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG1 |
| Immunohistochemistry | |
| 1001802 | |
| Immunohistochemistry 5-25 μg/mL | |
| Q6ZQW0 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human IDO2 in direct ELISAs. | |
| Human | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction