Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Iduronate 2-Sulfatase/IDS Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB2449
This item is not returnable.
View return policy
Description
Iduronate 2-Sulfatase/IDS Monoclonal specifically detects Iduronate 2-Sulfatase/IDS in Human samples. It is validated for Western Blot.Specifications
Iduronate 2-Sulfatase/IDS | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL | |
P22304 | |
IDS | |
Mouse myeloma cell line NS0-derived recombinant human Iduronate 2-Sulfatase/IDS Ser26-Pro550 Accession # P22304 | |
100 μg | |
Primary | |
Detects human Iduronate 2-Sulfatase/IDS in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody does not cross-react with recombinant mouse Iduronate 2-Sulfatase/IDS. | |
Human | |
Purified |
Western Blot | |
331320 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Alpha-L-iduronate sulfate sulfatase, EC 3.1.6.13, iduronate 2-sulfatase, iduronate 2-sulfatase 14 kDa chain, iduronate 2-sulfatase 42 kDa chain, idursulfase, MPS2, S, SIDS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
3423 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction