Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IFITM2/IFITM3 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF4834SP
This item is not returnable.
View return policy
Description
IFITM2/IFITM3 Polyclonal specifically detects IFITM2/IFITM3 in Human samples. It is validated for Western Blot.Specifications
IFITM2/IFITM3 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Goat | |
Affinity Purified | |
RUO | |
Detects human IFITM2 and human IFITM3 in direct ELISAs and Western blots. In direct ELISAs, less than 4% cross-reactivity with recombinant human IFITM1 is observed. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL | |
Q01628 | |
E. coli-derived human IFITM3 His3-His57 Accession # Q01628 | |
25 μg | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction