Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IFN-alpha 2/IFNA2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB93452100
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB93452100 100 μg
MAB93452SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB93452100 Supplier R&D Systems Supplier No. MAB93452100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

IFN-alpha 2 Monoclonal specifically detects IFN-alpha 2 in Human samples. It is validated for Neutralization.

Specifications

Antigen IFN-alpha 2
Applications Neutralization
Classification Monoclonal
Clone 967305
Conjugate Unconjugated
Dilution Neutralization 0.6-3.6 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. P01563
Gene Alias alpha-2a interferon, Ifa2, IFNA, IFNA2, IFNA2a, IFNA2b, IFNA2c, IFNAA, IFNalpha 2, IFN-alpha-2, IFN-alphaA, INFA2, interferon alpha 2b, interferon alpha A, interferon alpha-2, Interferon alpha-A, interferon, alpha 2, LeIF A, MGC125764, MGC125765
Host Species Mouse
Immunogen HEK293 human embryonic kidney cell line transfected with human IFN-alpha 2/IFNA2, Met1-Glu188, Accession # P01563
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 3440
Test Specificity Detects human IFN-alpha 2/IFNA2 in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.