Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IFN-alpha 2/IFNA2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $502.50
Specifications
Antigen | IFN-alpha 2 |
---|---|
Clone | 967314 |
Dilution | Immunocytochemistry 8-25 ug/mL |
Applications | Immunocytochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB9345SP
![]() |
R&D Systems
MAB9345SP |
25 μg |
Each for $200.50
|
|
|||||
MAB9345100
![]() |
R&D Systems
MAB9345100 |
100 μg |
Each for $502.50
|
|
|||||
Description
IFN-alpha 2 Monoclonal specifically detects IFN-alpha 2 in Human samples. It is validated for Immunocytochemistry.Specifications
IFN-alpha 2 | |
Immunocytochemistry 8-25 ug/mL | |
Monoclonal | |
Mouse | |
Human | |
P01563 | |
3440 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
967314 | |
Immunocytochemistry | |
Unconjugated | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
alpha-2a interferon, Ifa2, IFNA, IFNA2, IFNA2a, IFNA2b, IFNA2c, IFNAA, IFNalpha 2, IFN-alpha-2, IFN-alphaA, INFA2, interferon alpha 2b, interferon alpha A, interferon alpha-2, Interferon alpha-A, interferon, alpha 2, LeIF A, MGC125764, MGC125765 | |
Human embryonic kidney cell line HEK-293-derived transfected with human IFN-alpha 2/IFNA2, Met1-Glu188, Accession # P01563 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human IFN-alpha 2/IFNA2 in ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) IFNE, rhIFNB, rhIFNG, or recombinant cotton rat IFNA is observed. In sandwich ELISAs, less than 8% cross-reactivity with rhIFNA-7, rhIFNA-8, and rhIFNA-10 is observed. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title