Learn More
R&D Systems™ Human IFN-alpha All Subtype Quantikine ELISA Kit

Specifications
Specifications
| Assay Range | 15.60 to 1000 pg/mL |
| Assay Sensitivity | 4.8 pg/mL |
| Sample Type | Cell Culture Supernatant, Plasma (EDTA), Plasma (Heparin) |
| Specificity | Natual and recombinant human IFN-alpha . |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 3440 |
| Quantity | 1 Kit |
| Storage Requirements | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Synonym | IFNA, IFNA2, IFNA2b, IFNalpha, IFN-alphaA, INFA2, interferon alpha 2, LeIF D |
| Target | Human |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.