Learn More
Human IFN-alpha Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | IFN-alpha |
| Applications | Neutralization |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Neutralization |
| Formulation | Neat serum diluted in phosphate buffered saline (PBS) |
| Gene Alias | IFNA, IFNA2, IFNA2b, IFNalpha, IFN-alphaA, INFA2, interferon alpha 2, LeIF D |
| Host Species | Sheep |
| Immunogen | Mixture of all recombinant human interferon alpha subtypes (IFNA2, IFNA8, IFNA10, IFNA1, IFNA21, IFNA5, IFNA14, IFNA17, IFNA7, IFNA6, IFNA4, IFNA16) |
| Quantity | 100,000 U |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.