Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IFN-alpha Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
Supplier: R&D Systems 211121
This item is not returnable.
View return policy
Description
IFN-alpha Monoclonal specifically detects IFN-alpha in Human samples. It is validated for ELISA, Neutralization.Specifications
IFN-alpha | |
Monoclonal | |
Unconjugated | |
Phosphate-buffered saline (PBS) containing 0.1% bovine serum albumin (BSA) with No Preservative | |
IFNA2 | |
Human Leukocyte Interferon | |
50 μg | |
Primary | |
One neutralization unit is the amount of antiserum required to neutralize one unit of human interferon alpha (Hu-IFN-alpha ) to a 50% endpoint. Interferon was titrated with the use of the cytopathic effect inhibition assay as described [Rubinstein, S., Familletti, P.C., and Pestka, S. (1981) “Convenient Assay for Interferons,” J. Virol . 37, 755-758; Familletti, P.C., Rubinstein, S., and Pestka, S. (1981) “A Convenient and Rapid Cytopathic Effect Inhibition Assay for Interferon,” in Methods in Enzymology, Vol. 78 (S. Pestka, ed.), Academic Press, New York, 387-394]. In this antiviral assay for interferon, about 1 unit/ml of interferon is the quantity necessary to produce a cytopathic effect of 50%. The units are determined with respect to the international reference standard for Hu-IFN-alpha provided by the National Institutes of Health [see Pestka, S. (1986) “Interferon Standards and General Abbreviations,” in Methods in Enzymology (S. Pestka, ed.), Academic Press, New York 119, 14-23]. | |
After receipt, the product may be stored at -20degreesC for short-term use ( < 6 months). For long-term storage, we recommend storing the product at -70degreesC or below for retention of full activity. When thawing, the contents of the tube should be apportioned in separate tubes so that freezing and thawing is kept to a minimum. Refreezing should be done on dry ice or in a dry ice/alcohol bath. Further dilution of the product should be in buffers containing protein such as 0.1% bovine serum albumin (BSA). | |
IgG1 |
ELISA, Neutralization | |
MMHA-11 | |
ELISA, Neutralization | |
IFNA, IFNA2, IFNA2b, IFNalpha, IFN-alphaA, INFA2, interferon alpha 2, LeIF D | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
3440 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction