Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IFN-alpha Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
Supplier: R&D Systems 211161
This item is not returnable.
View return policy
Description
IFN-alpha Monoclonal specifically detects IFN-alpha in Human samples. It is validated for Direct ELISA.Specifications
IFN-alpha | |
Monoclonal | |
Unconjugated | |
Phosphate-buffered saline (PBS) containing 0.1% bovine serum albumin (BSA) with No Preservative | |
IFNA2 | |
Human Leukocyte Interferon | |
50 μg | |
Primary | |
Detects human interferon alpha in direct ELISAs. | |
After receipt, the product may be stored at -20degreesC for short-term use ( < 6 months). For long-term storage, we recommend storing the product at -70degreesC or below for retention of full activity. When thawing, the contents of the tube should be apportioned in separate tubes so that freezing and thawing is kept to a minimum. Refreezing should be done on dry ice or in a dry ice/alcohol bath. Further dilution of the product should be in buffers containing protein such as 0.1% bovine serum albumin (BSA). | |
IgG1 |
ELISA | |
MMHA-13 | |
Direct ELISA | |
IFNA, IFNA2, IFNA2b, IFNalpha, IFN-alphaA, INFA2, interferon alpha 2, LeIF D | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
3440 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction