Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IFN-alpha Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. 211181
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
211181 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 211181 Supplier R&D Systems Supplier No. 211181
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

IFN-alpha Monoclonal specifically detects IFN-alpha in Human samples. It is validated for ELISA, Neutralization.

Specifications

Antigen IFN-alpha
Applications ELISA, Neutralization
Classification Monoclonal
Clone MMHA-17
Conjugate Unconjugated
Dilution ELISA, Neutralization
Formulation Phosphate-buffered saline (PBS) containing 0.1% bovine serum albumin (BSA) with No Preservative
Gene Alias IFNA, IFNA2, IFNA2b, IFNalpha, IFN-alphaA, INFA2, interferon alpha 2, LeIF D
Gene Symbols IFNA2
Host Species Mouse
Immunogen Human Leukocyte Interferon
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3440
Test Specificity Neutralizes human interferon alpha; binds to human interferon alpha
Target Species Human
Content And Storage After receipt, the product may be stored at -20degreesC for short-term use ( < 6 months). For long-term storage, we recommend storing the product at -70degreesC or below for retention of full activity. When thawing, the contents of the tube should be apportioned in separate tubes so that freezing and thawing is kept to a minimum. Refreezing should be done on dry ice or in a dry ice/alcohol bath. Further dilution of the product should be in buffers containing protein such as 0.1% bovine serum albumin (BSA).
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.