Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IFN-alpha/beta R1 Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems BAF245
This item is not returnable.
View return policy
Description
IFN-alpha/beta R1 Polyclonal specifically detects IFN-alpha/beta R1 in Human samples. It is validated for Western Blot, Flow Cytometry.Specifications
IFN-alpha/beta R1 | |
Polyclonal | |
Biotin | |
AAA52730 | |
IFNAR1 | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IFN-alpha/beta R1 Lys28-Lys436 Accession # AAA52730 | |
50 μg | |
Primary | |
Detects human IFN-alpha/beta R1 in Western blots. | |
Human | |
IgG |
Western Blot, Flow Cytometry | |
LYOPH | |
Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells | |
alpha-type antiviral protein, AVP, beta-type antiviral protein, CRF2-1, Cytokine receptor class-II member 1, Cytokine receptor family 2 member 1, human interferon-alpha receptor (HuIFN-alpha-Rec)10IFRC, IFN-alpha/beta receptor 1, IFN-alpha-REC, IFNAR, IFN-aR1, IFNAR1, IFNBR, IFNbR1, IFN-bR1, IFN-R-1, interferon (alpha, beta and omega) receptor 1, interferon alpha/beta receptor 1, interferon-alpha/beta receptor alpha chain, interferon-beta receptor 1, Type I interferon receptor 1 | |
Goat | |
Affinity Purified | |
RUO | |
3454 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction