Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IFN-alpha/beta R2 C-Terminus Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF7014
This item is not returnable.
View return policy
Description
IFN-alpha/beta R2 Polyclonal specifically detects IFN-alpha/beta R2 in Human samples. It is validated for Western Blot.Specifications
IFN-alpha/beta R2 | |
Polyclonal | |
Unconjugated | |
P48551 | |
IFNAR2 | |
E. coli-derived recombinant human IFN-alpha/beta R2 C-Terminus Ser351-Met514 Accession # P48551 | |
100 μg | |
Primary | |
Detects human IFN-alpha/beta R2 C-Terminus in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) IFN-alpha/beta R2a, rhIFN-alpha/beta R alpha , and recombinant mouse IFN-alpha/beta R2 is observed. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL | |
beta receptor10, human interferon alpha, human interferon alpha/beta receptor, IFNABR, IFN-alpha binding protein, IFN-alpha/beta receptor 2, IFN-alpha-REC, IFN-aR2, IFNAR2, IFNARB, IFNbR2, IFN-bR2, IFN-R, IFN-R-2, interferon (alpha, beta and omega) receptor 2, Interferon alpha binding protein, interferon alpha/beta receptor 2, interferon-alpha/beta receptor beta chain, Type I interferon receptor 2 | |
Sheep | |
Affinity Purified | |
RUO | |
3455 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction