Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IFN-alpha I Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB100801SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB100801SP 25 μg
MB100801100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB100801SP Supplier R&D Systems Supplier No. MAB100801SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

IFN-alpha I/IFNA17 Monoclonal specifically detects IFN-alpha I/IFNA17 in Human samples. It is validated for Neutralization.

Specifications

Antigen IFN-alpha I/IFNA17
Applications Neutralization
Classification Monoclonal
Clone 995819
Conjugate Unconjugated
Dilution Neutralization 6-36 ng/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. P01571
Gene Alias IFNA, IFNA17, IFNalpha I, IFN-alpha-17, IFN-alphaI, INFA, interferon alpha-17, Interferon alpha-88, Interferon alpha-I', Interferon alpha-T, interferon alpha-WA, interferon, alpha 17, LeIF I, LEIF2C1
Host Species Mouse
Immunogen Human embryonic kidney cell line HEK293-derived recombinant human IFN-alpha I, Met1-Asp189, Accession # P01571
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Primary or Secondary Primary
Gene ID (Entrez) 3451
Test Specificity Detects human IFN-alpha I in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.