Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human IFN-alpha Serum Sample ELISA Kit

Catalog No. 411101
Click to view available options
Quantity:
1 Plate
5 Plates
This item is not returnable. View return policy
This item is not returnable. View return policy

Human IFN-alpha Serum Sample ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human IFN-alpha Serum Sample ELISA Kit quantifies human IFN-alpha in cell culture supernates (100 ul), serum (100 ul), plasma (100 ul).

Specifications

Assay Sensitivity 12.5pg/mL
Conjugate Biotin
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Type Solid Phase Sandwich ELISA
Sample Type Cell Culture Supernatant, Plasma, Serum
Specificity Human IFN-alpha
Sufficient For 1 x 96-well Plate
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 3440
Quantity 1 Plate
Sample Volume Cell Culture Supernates (100 μL), Serum (100 μL), Plasma (100 μL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym IFNA, IFNA2, IFNA2b, IFNalpha, IFN-alphaA, INFA2, interferon alpha 2, LeIF D
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.