Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IFN-beta Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. 214651
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
214651 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 214651 Supplier R&D Systems Supplier No. 214651
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

IFN-beta Monoclonal specifically detects IFN-beta in Human samples. It is validated for Direct ELISA.

Specifications

Antigen IFN-beta
Applications ELISA
Classification Monoclonal
Clone MMHB-15
Conjugate Unconjugated
Dilution Direct ELISA
Formulation Phosphate buffered saline (PBS) containing 0.1% bovine serum albumin (BSA) with No Preservative
Gene Alias Fibroblast interferon, IFB, IFBIFNB, IFF, IFNB, IFNB1, IFNbeta, IFN-beta, interferon beta, interferon, beta 1, fibroblast, MGC96956
Gene Symbols IFNB1
Host Species Mouse
Immunogen Human interferon beta
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3456
Test Specificity Binds to human interferon beta with high affinity. The ND 50 for this antibody on human (A549/EMCV) cells is ∼27-66 μg/ml in the presence of 96 pg/ml of recombinant human interferon beta 1a, mammalian.
Target Species Human
Content And Storage After receipt, the product may be stored at -20degreesC for short-term use (< 6 months). For long-term storage, we recommend storing the product at -70degreesC or below for retention of full activity. When thawing, the contents of the tube should be apportioned in separate tubes so that freezing and thawing is kept to a minimum. Refreezing should be done on dry ice or in a dry ice/alcohol bath. Further dilution of the product should be in buffers containing protein such as 0.1% bovine serum albumin (BSA).
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.