Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IFN-beta Antibody, R&D Systems™

Mouse Monoclonal Antibody
$176.48 - $532.33
Specifications
| Antigen | IFN-beta |
|---|---|
| Clone | 76703R |
| Dilution | Neutralization 7-21 ug/mL |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB814RSP
![]() |
R&D Systems
MAB814RSP |
25 μg |
Each for $176.48
|
|
|||||
MAB814R100
![]() |
R&D Systems
MAB814R100 |
100 μg |
Each for $532.33
|
|
|||||
Description
IFN-beta Monoclonal specifically detects IFN-beta in Human samples. It is validated for Neutralization.Specifications
| IFN-beta | |
| Neutralization 7-21 ug/mL | |
| Unconjugated | |
| Mouse | |
| Human | |
| Fibroblast interferon, IFB, IFBIFNB, IFF, IFNB, IFNB1, IFNbeta, IFN-beta, interferon beta, interferon, beta 1, fibroblast, MGC96956 | |
| IFNB1 | |
| Primary | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 76703R | |
| Monoclonal | |
| Purified | |
| RUO | |
| P01574 | |
| 3456 | |
| IgG1 | |
| Protein A or G purified from cell culture supernatant | |
| Detects human IFN-beta in direct ELISAs. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title