Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IFN-epsilon Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB91471SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB91471SP 25 μg
MAB91471100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB91471SP Supplier R&D Systems Supplier No. MAB91471SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

IFN-epsilon Monoclonal specifically detects IFN-epsilon in Human samples. It is validated for ELISA.

Specifications

Antigen IFN-epsilon
Applications ELISA
Classification Monoclonal
Clone 983303
Conjugate Unconjugated
Dilution ELISA
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. Q86WN2
Gene Alias IFNE, IFN-E, IFNE1PRO655, IFNepsilon, IFN-epsilon, IFNT1, Ifn-tau-11, INFE1, interferon epsilon, interferon epsilon 1, Interferon epsilon-1, interferon tau-1, interferon, epsilon, interferon-epsilon, MGC119018, MGC119020
Host Species Mouse
Immunogen E. coli-derived recombinant human IFN-epsilon, Leu22-Arg208, Accession No. Q86WN2
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Primary or Secondary Primary
Gene ID (Entrez) 338376.0
Test Specificity Detects human IFN-epsilon in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Form Lyophilized
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.