Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IFN-epsilon Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $514.50
Specifications
Antigen | IFN-epsilon |
---|---|
Clone | 983338 |
Dilution | Immunohistochemistry 1-25 ug/mL |
Applications | Immunohistochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB9147SP
![]() |
R&D Systems
MAB9147SP |
25 μg |
Each for $200.50
|
|
|||||
MAB9147100
![]() |
R&D Systems
MAB9147100 |
100 μg |
Each for $514.50
|
|
|||||
Description
IFN-epsilon Monoclonal specifically detects IFN-epsilon in Human samples. It is validated for ELISA, Immunohistochemistry.Specifications
IFN-epsilon | |
Immunohistochemistry 1-25 ug/mL | |
Monoclonal | |
Supernatant | |
Human | |
Q86WN2 | |
338376 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
983338 | |
Immunohistochemistry | |
Unconjugated | |
Mouse | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
IFNE, IFN-E, IFNE1PRO655, IFNepsilon, IFN-epsilon, IFNT1, Ifn-tau-11, INFE1, interferon epsilon, interferon epsilon 1, Interferon epsilon-1, interferon tau-1, interferon, epsilon, interferon-epsilon, MGC119018, MGC119020 | |
E. coli-derived recombinant human IFN-epsilon Leu22-Arg208 Accession # Q86WN2 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human IFN-epsilon in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title