Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IFN-gamma Alexa Fluor 405-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. IC2852V100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC2852V100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC2852V100 Supplier R&D Systems Supplier No. IC2852V100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

IFN-gamma Monoclonal specifically detects IFN-gamma in Human samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen IFN-gamma
Applications Flow Cytometry
Classification Monoclonal
Clone K3.53
Conjugate Alexa Fluor 405
Dilution Intracellular Staining by Flow Cytometry 0.25-1 μg/106 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Alias IFG, IFI, IFNG, IFNgamma, IFN-gamma, Immune interferon, interferon gamma, interferon, gamma
Host Species Mouse
Immunogen E. coli-derived recombinant human IFN-gamma
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 3458.0
Test Specificity Detects human IFN-gamma in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant mouse IFN-gamma , recombinant rat IFN-gamma , recombinant porcine IFN-gamma , recombinant feline IFN-gamma , or recombinant cotton rat IFN-gamma is observed.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Form Supernatant
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.