Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IFN-gamma Alexa Fluor 405-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems IC2851V025
This item is not returnable.
View return policy
Description
IFN-gamma Monoclonal specifically detects IFN-gamma in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
IFN-gamma | |
Monoclonal | |
Alexa Fluor 405 | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
IFG, IFI, IFNG, IFNgamma, IFN-gamma, Immune interferon, interferon gamma, interferon, gamma | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
3458 | |
Human | |
Purified |
Flow Cytometry | |
25718 | |
Intracellular Staining by Flow Cytometry 5 uL/10^6 cells | |
AAP20098.1 | |
IFNG | |
E. coli-derived recombinant human IFN-gamma Gln24-Gln166 Accession # AAP20098.1 | |
25 Tests | |
Primary | |
Detects human IFN-gamma in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse IFN-gamma , recombinant rat IFN-gamma , or recombinant porcine IFN-gamma is observed. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction