Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IFN-gamma Antibody, R&D Systems™
GSA_VAGREENER_CHOICE

Mouse Monoclonal Antibody has been used in 15 publications

$241.00 - $2070.00

Specifications

Antigen IFN-gamma
Clone K3.53
Dilution Western Blot 1 ug/mL, Neutralization ≤2 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Applications Western Blot, Neutralization, ELISA
Classification Monoclonal
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB2852SP
GREENER_CHOICE
View Documents
R&D Systems
MAB2852SP
25 μg
Each for $241.00
Only null left
Add to Cart
 
MAB2852100
GREENER_CHOICE
View Documents
R&D Systems
MAB2852100
100 μg
Each for $561.50
Only null left
Add to Cart
 
MAB2852500
GSA_VAGREENER_CHOICE
View Documents
R&D Systems
MAB2852500
500 μg
Each for $2,070.00
Only null left
Add to Cart
 
Description

Description

IFN-gamma Monoclonal specifically detects IFN-gamma in Human samples. It is validated for Western Blot, Neutralization, Intracellular Staining by Flow Cytometry, ELISA Capture (Matched Antibody Pair).
Specifications

Specifications

IFN-gamma
Western Blot 1 ug/mL, Neutralization ≤2 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Monoclonal
Mouse
Human
IFG, IFI, IFNG, IFNgamma, IFN-gamma, Immune interferon, interferon gamma, interferon, gamma
IFNG
IgG2a
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
K3.53
Western Blot, Neutralization, ELISA
Unconjugated
RUO
Lyophilized from a 0.2 μm filtered solution in PBS and NaCl with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
3458
E. coli-derived recombinant human IFN-gamma
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects human IFN-gamma in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant mouse IFN-gamma , recombinant rat IFN-gamma , recombinant porcine IFN-gamma , recombinant feline IFN-gamma , or recombinant cotton rat IFN-gamma is observed.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.