Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IFN-gamma Antibody, R&D Systems™



Mouse Monoclonal Antibody
$241.00 - $2150.00
Specifications
Antigen | IFN-gamma |
---|---|
Clone | 25718 |
Dilution | Neutralization 0.02-0.06 ug/mL, Immunocytochemistry 8-25 ug/mL |
Applications | Neutralization, Immunocytochemistry |
Classification | Monoclonal |
Description
IFN-gamma Monoclonal specifically detects IFN-gamma in Human samples. It is validated for Neutralization, Intracellular Staining by Flow Cytometry, Immunocytochemistry.Specifications
IFN-gamma | |
Neutralization 0.02-0.06 ug/mL, Immunocytochemistry 8-25 ug/mL | |
Monoclonal | |
Mouse | |
Human | |
AAP20098.1 | |
3458 | |
E. coli-derived recombinant human IFN-gamma Gln24-Gln166 Accession # AAP20098.1 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human IFN-gamma . |
25718 | |
Neutralization, Immunocytochemistry | |
Unconjugated | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
IFG, IFI, IFNG, IFNgamma, IFN-gamma, Immune interferon, interferon gamma, interferon, gamma | |
IFNG | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title