Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IFN-gamma Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB285RSP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB285R-SP 25 μg
MAB285R-100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB285R-SP Supplier R&D Systems Supplier No. MAB285RSP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

IFN-gamma Monoclonal specifically detects IFN-gamma in Human samples. It is validated for Neutralization, Immunocytochemistry.

Specifications

Antigen IFN-gamma
Applications Immunocytochemistry, Neutralization
Classification Monoclonal
Clone 25718R
Conjugate Unconjugated
Dilution Neutralization 0.02-0.06μg/mL, Immunocytochemistry 8-25μg/mL
Formulation Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS.
Gene Accession No. AAP20098.1
Gene Alias IFG, IFI, IFNG, IFNgamma, IFN-gamma, Immune interferon, interferon gamma, interferon, gamma
Host Species Mouse
Immunogen E. coli-derived recombinant human IFN-gamma Gln24-Gln166 Accession # AAP20098.1
Purification Method Protein A or G purified from cell culture supernatant
Quantity 25 μg
Primary or Secondary Primary
Gene ID (Entrez) 3458
Test Specificity Detects human IFN-gamma in direct ELISA.
Reconstitution Reconstitute at 0.5mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.