Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IFN-gamma Fluorescein-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 8 publications
$362.50
Specifications
Antigen | IFN-gamma |
---|---|
Clone | 25723 |
Dilution | Intracellular Staining by Flow Cytometry 10 uL/10^6 cells |
Applications | Flow Cytometry |
Classification | Monoclonal |
Description
IFN-gamma Monoclonal specifically detects IFN-gamma in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
IFN-gamma | |
Intracellular Staining by Flow Cytometry 10 uL/10^6 cells | |
Monoclonal | |
Purified | |
RUO | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
IFG, IFI, IFNG, IFNgamma, IFN-gamma, Immune interferon, interferon gamma, interferon, gamma | |
IFNG | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Detects human IFN-gamma in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse IFN-gamma , recombinant rat IFN-gamma , or recombinant porcine IFN-gamma is observed. |
25723 | |
Flow Cytometry | |
Fluorescein | |
Mouse | |
Human | |
AAP20098.1 | |
3458 | |
E. coli-derived recombinant human IFN-gamma Gln24-Gln166 Accession # AAP20098.1 | |
Primary | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title