Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IFN-gamma PerCP-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. IC285C100
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC285C100 100 Tests
IC285C025 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. IC285C100 Supplier R&D Systems Supplier No. IC285C100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

IFN-gamma Monoclonal specifically detects IFN-gamma in Human samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen IFN-gamma
Applications Flow Cytometry
Classification Monoclonal
Clone 25723
Conjugate PerCP
Dilution Intracellular Staining by Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. AAP20098.1
Gene Alias IFG, IFI, IFNG, IFNgamma, IFN-gamma, Immune interferon, interferon gamma, interferon, gamma
Gene Symbols IFNG
Host Species Mouse
Immunogen E. coli-derived recombinant human IFN-gamma Gln24-Gln166 Accession # AAP20098.1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3458
Test Specificity Detects human IFN-gamma in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse IFN-gamma , recombinant rat IFN-gamma , or recombinant porcine IFN-gamma is observed.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.