Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IFN-gamma R1/CD119 Alexa Fluor 405-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB6731V100UG
This item is not returnable.
View return policy
Description
IFN-gamma R1/CD119 Monoclonal specifically detects IFN-gamma R1/CD119 in Human samples. It is validated for Flow Cytometry.Specifications
IFN-gamma R1/CD119 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
P15260.1 | |
IFNGR1 | |
Mouse myeloma cell line NS0-derived recombinant human IFN-gamma R1 Glu18-Gly245 Accession # P15260.1 | |
RUO | |
3459 | |
Human | |
IgG1 |
Flow Cytometry | |
92101 | |
Alexa Fluor 405 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
AVP, type 2, CD119, CD119 antigen, FLJ45734, IFN-gamma receptor 1, IFNgammaR1, IFN-gamma-R1, IFNGR, IFN-gR1, IFNGR1, immune interferon receptor 1, interferon gamma receptor 1, interferon-gamma receptor alpha chain, type 2 | |
Mouse | |
100 μg | |
Primary | |
Detects human IFN-gamma R1 in direct ELISAs and Western blots. In direct ELISAs, this antibody does not cross-react with recombinant mouse IFN-gamma R1, recombinant human (rh) IFN-gamma R2, or rhIL-10 R beta . | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction