Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IFN-gamma R2 Fluorescein-conjugated Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems FAB773F
This item is not returnable.
View return policy
Description
IFN-gamma R2 Polyclonal specifically detects IFN-gamma R2 in Human samples. It is validated for Flow Cytometry.Specifications
IFN-gamma R2 | |
Polyclonal | |
Flow Cytometry 10 uL/10^6 cells | |
AF-1, AF-1interferon-gamma receptor beta chain precursor10IFNGT1Interferon gamma receptor accessory factor 1, IFGR2, IFN-gamma receptor 2, IFNgammaR2, IFN-gamma-R2, IFN-gR2, IFNGR2, IFNGT1, interferon gamma receptor 2, interferon gamma receptor 2 (interferon gamma transducer 1), interferon gamma receptor accessory factor-1, interferon gamma receptor beta chain, Interferon gamma transducer 1 | |
Goat | |
Affinity Purified | |
RUO | |
3460 | |
Human | |
IgG |
Flow Cytometry | |
Fluorescein | |
P38484 | |
IFNGR2 | |
Mouse myeloma cell line NS0-derived recombinant human IFN-gamma R2 Ala22-Gln247 Accession # P38484 | |
100 Tests | |
Primary | |
Detects human IFN-gamma R2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human IFN-gamma R1 is observed. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction