Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human IFN-lambda 4 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB9165100
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB9165-100 100 μg
MAB9165-SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB9165-100 Supplier R&D Systems Supplier No. MAB9165100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

IFN-lambda 4 Monoclonal specifically detects IFN-lambda 4 in Human samples. It is validated for Immunohistochemistry.

Specifications

Antigen IFN-lambda4
Applications Immunohistochemistry
Classification Monoclonal
Clone 991620
Conjugate Unconjugated
Dilution Immunohistochemistry 5-25μg/mL
Formulation Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS.
Gene Accession No. K9M1U5
Gene Alias IFNAN, IFNL4, IFNlambda 4
Host Species Mouse
Immunogen E. coli-derived recombinant human IFN-lambda 4 Ala23-Leu179 Accession # K9M1U5
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 101180976
Test Specificity Detects human IFN-lambda 4 in direct ELISAs.
Reconstitution Reconstitute at 0.5mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.