Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Ig Kappa Light Chain Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $502.50
Specifications
Antigen | Human Kappa Light Chain |
---|---|
Clone | 999947 |
Dilution | Western Blot 2 μg/mL, ELISA |
Applications | Western Blot, ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB100501SP
![]() |
R&D Systems
MAB100501SP |
25 μg |
Each for $200.50
|
|
|||||
MB100501100
![]() |
R&D Systems
MAB100501100 |
100 μg |
Each for $502.50
|
|
|||||
Description
Human Kappa Light Chain Monoclonal specifically detects Human Kappa Light Chain in Human samples. It is validated for Western Blot, Simple Western, ELISA.Specifications
Human Kappa Light Chain | |
Western Blot 2 μg/mL, ELISA | |
Monoclonal | |
Supernatant | |
Human | |
Human Ig Kappa light chain, | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects Human Ig Kappa light chain in direct ELISAs. |
999947 | |
Western Blot, ELISA | |
Unconjugated | |
Mouse | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title