Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Ig lambda C Domain Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF6525SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF6525SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF6525SP Supplier R&D Systems Supplier No. AF6525SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Ig Lambda C Domain/IGLC2 Polyclonal specifically detects Ig Lambda C Domain/IGLC2 in Human samples. It is validated for Western Blot.

Specifications

Antigen Ig Lambda C Domain/IGLC2
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P0CG05
Gene Alias IGLC, IGLC2, immunoglobulin lambda constant 2 (Kern-Oz- marker), MGC20392, MGC45681
Gene Symbols IGLC2
Host Species Goat
Immunogen E. coli-derived recombinant human IGLC2 Gln2-Ser106 Accession # P0CG05
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3538
Test Specificity Detects recombinant human IGLC2 in direct ELISAs and Western blots. Detects endogenous Immunoglobulin lambda constant region in natural samples by Western blots.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.