Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Ig Lambda Light Chain Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MB100491100
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MB100491100 100 μg
MAB100491SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MB100491100 Supplier R&D Systems Supplier No. MAB100491100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Human lambda light chain Monoclonal specifically detects Human lambda light chain in Human samples. It is validated for Western Blot, Simple Western, ELISA.

Specifications

Antigen Human lambda light chain
Applications Western Blot, ELISA
Classification Monoclonal
Clone 1000040
Conjugate Unconjugated
Dilution Western Blot 2 μg/mL, Simple Western 20 μg/mL, ELISA
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Alias Bence Jones protein, BJP, Constant region of lambda light chains, HCAK 1, HCAK1, Ig kappa chain C region, Ig lambda chain C regions, IGKC, IGLC, IGLC 1, IGLC 2, IGLC 3, IGLC1, IGLC2, IGLC3, Immunoglobulin kappa constant, Immunoglobulin kappa constant region, Immunoglobulin kappa light chain, Immunoglobulin lambda constant 1, Kappa 1 immunoglobulin light chain, Km, Mcg marker, MGC111575, MGC62011, MGC72072, MGC88770, MGC88771, MGC88809, Paraprotein
Host Species Mouse
Immunogen Ig Lamda light chain,
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Primary or Secondary Primary
Test Specificity Detects Human Ig Lambda Light Chain in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.