Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Ig lambda light chain Rabbit anti-Human, Clone: RM127 (Biotin), Abnova™

Catalog No. 89035302 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-035-302 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-035-302 Supplier Abnova Corporation Supplier No. MAB14950
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Rabbit monoclonal antibody raised against native human Ig lambda light chain.

  • High specificity
  • High reactivity

Specifications

Antigen Human Ig lambda light chain
Applications ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (PFA fixed)
Classification Monoclonal
Clone RM127
Conjugate Biotin
Description Rabbit monoclonal antibody raised against native human Ig lambda light chain.
Dilution ELISA (0.05-0.2 ug/mL) Flow Cytometry Immunocytochemistry (0.5-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-2 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (50% glycerol, 1% BSA, 0.09% sodium azide)
Gene Accession No. P0CG04
Gene Alias IGLC
Gene Symbols IGLC1
Host Species Rabbit
Immunogen Native purified human IgG.
Purification Method Protein A purification
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3537
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.